Crocein Scarlet 7B, >70% dye content
Crocein Scarlet 7B, >70% dye content
Crocein Scarlet 7B, >70% dye content
Crocein Scarlet 7B, >70% dye content
Thermo Scientific Chemicals

Crocein Scarlet 7B, >70% dye content

CAS: 6226-76-2 | C24H18N4Na2O7S2
Have Questions?
Change viewbuttonViewtableView
Quantity:
1g
5g
25g
Catalog number J66876.03
also known as J66876-03
Price (EUR)
25,60
Each
Quantity:
1g
Request bulk or custom format
Price (EUR)
25,60
Each
Specifications
CAS6226-76-2
ColorRed
FormPowder
FormatPowder
Formula Weight584.53
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only