Ceftiofur sodium, 95%
Ceftiofur sodium, 95%
Ceftiofur sodium, 95%
Thermo Scientific™

Ceftiofur sodium, 95%

A broad spectrum antibiotic | CAS: 104010-37-9 | C19H16N5NaO7S3 | 545.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66886.03
also known as J66886-03
Price (EUR)
71,30
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
71,30
Each
Specifications
CAS104010-37-9
Chemical Name or MaterialCeftiofur sodium
Formula Weight545.54
Health Hazard 1H317-H334
Health Hazard 2GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
View more
Ceftiofur sodium is resistant to the antibiotic resistance enzyme beta-lactamase, and has activity against Gram-positive and Gram-negative bacteria. E. coli strains resistant to ceftiofur have been reported. The bactericidal activity of ceftiofur results from the inhibition of cell wall synthesis via affinity for penicillin-binding proteins. The PBPs are transpeptidases which are vital in peptidoglycan biosynthesis. Therefore, their inhibition prevents this vital cell wall component from being properly synthesized.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ceftiofur sodium is resistant to the antibiotic resistance enzyme beta-lactamase, and has activity against Gram-positive and Gram-negative bacteria. E. coli strains resistant to ceftiofur have been reported. The bactericidal activity of ceftiofur results from the inhibition of cell wall synthesis via affinity for penicillin-binding proteins. The PBPs are transpeptidases which are vital in peptidoglycan biosynthesis. Therefore, their inhibition prevents this vital cell wall component from being properly synthesized.

Solubility
Freely soluble in aqueous solution

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only
Beate G. Rose.; Carol Kamps-Holtzapple.; Larry H. Stanker. Competitive Indirect ELISA for Ceftiofur Sodium and the Effect of Different Immunizing and Coating Antigen Conjugates. Bioconjugate Chem. 1995, 6, (5), 529-535.