1,2,3,5-Tetra-O-benzoyl-2-C-methyl-beta-D-ribofuranose, 98%, Thermo Scientific Chemicals
1,2,3,5-Tetra-O-benzoyl-2-C-methyl-beta-D-ribofuranose, 98%, Thermo Scientific Chemicals
1,2,3,5-Tetra-O-benzoyl-2-C-methyl-beta-D-ribofuranose, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2,3,5-Tetra-O-benzoyl-2-C-methyl-beta-D-ribofuranose, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
J66930.06
also known as J66930-06
5 g
Catalog number J66930.06
also known as J66930-06
Price (EUR)
125,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
125,00
Each
Specifications
CAS15397-15-6
Chemical Name or Material1,2,3,5-Tetra-O-benzoyl-2-C-methyl-beta-D-ribofuranose
ColorWhite
Formula Weight580.58
IUPAC Name[(2R,3R,4R,5S)-3,4,5-tribenzoyloxy-4-methyloxolan-2-yl]methyl benzoate
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only