Doxycycline hydrochloride, 20 mg/ml in distilled water, sterile-filtered
Doxycycline hydrochloride, 20 mg/ml in distilled water, sterile-filtered
Doxycycline hydrochloride, 20 mg/ml in distilled water, sterile-filtered
Thermo Scientific Chemicals

Doxycycline hydrochloride, 20 mg/ml in distilled water, sterile-filtered

Doxycycline hydrochloride, CAS # 10592-13-9, is the hydrochloride salt form of doxycycline that shows antibacterial and antimalarial activities. | CAS: 10592-13-9 | C22H25ClN2O8 | 480.90 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J67043.AD
also known as J67043-AD
Price (EUR)
138,00
Each
Quantity:
50 mL
Request bulk or custom format
Price (EUR)
138,00
Each
Chemical Identifiers
CAS10592-13-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow
FormLiquid
CommentComponent is dissolved in UV treated 18.2 megohm-cm ± 1, endotoxin tested water (Endotoxin content: ≥ 0.1EU/ml). The solution is filtered through 0.22 micron filter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

• Doxycycline hydrochloride is a compound that shows antibacterial and antimicrobial activities against several species, including Pseudomonas aeruginosa, Plasmodium falciparum, and Escherichia coli. It is also a bacteriostatic

• It can block the binding of aminoacyl-tRNA to the mRNA-ribosome complex, thereby inhibiting protein synthesis

Application

• It is used to eliminate Borrelia burgdorferi and Anaplasma phagocytophilum in rodent reservoirs and eliminate Ixodes scapularis ticks

• Doxycycline hydrochloride is also considered to be a broad-spectrum inhibitor used to inhibit matrix metalloproteinases (MMP), such as type 1 collagenase, in wound healing and tissue remodeling studies. Due to this activity, it can inhibit angiogenesis and reduce lung metastases

RUO – Research Use Only

General References:

  1. Z Qiang.; C Adams. Potentiometric determination of acid dissociation constants (pK a) for human and veterinary antibiotics. Water research. 2004, 38, (12), 2874-2890.
  2. JB Bogardus.; RK Blackwood. Solubility of doxycycline in aqueous solution. Journal of Pharmaceutical Sciences. 1979, 68, (2), 188-194.