Krebs-Ringer Solution, HEPES-buffered
Thermo Scientific Chemicals

Krebs-Ringer Solution, HEPES-buffered

Standard physiological buffer that is suitable for various calcium imaging experiments. It is also used as a preserving agent.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number J67795.K2
also known as J67795-K2
Price (EUR)
185,65
Online Exclusive
206,00
Save 20,35 (10%)
Each
Quantity:
1 L
Request bulk or custom format
Price (EUR)
185,65
Online Exclusive
206,00
Save 20,35 (10%)
Each
Specifications
Chemical Name or MaterialKrebs-Ringer Solution
PackagingPlastic bottle
Recommended StorageAmbient temperatures
Solubility InformationNot miscible or difficult to mix in water.
TSCAYes
View more
Standard physiological buffer that is suitable for various calcium imaging experiments. It is also used as a preserving agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only