Thermo Scientific Chemicals

3-Methyl-2,4-pentanedione, mixture of tautomers, 95%, Thermo Scientific Chemicals

Catalog number: L01135.14
25 g, Each
Thermo Scientific Chemicals

3-Methyl-2,4-pentanedione, mixture of tautomers, 95%, Thermo Scientific Chemicals

Catalog number: L01135.14
25 g, Each
Quantity
Catalog number: L01135.14
also known as L01135-14
Price (EUR)
Price: 115,00
Online price: 103,50
Your price:
Quantity
-

Chemical Identifiers

CAS
815-57-6
IUPAC Name
3-methylpentane-2,4-dione
Molecular Formula
C6H10O2
InChI Key
GSOHKPVFCOWKPU-UHFFFAOYSA-N
SMILES
CC(C(C)=O)C(C)=O
Form
liquid
Refractive Index
1.4410-1.4470
Appearance (Color)
Clear, colourless to yellow
Assay (GC)
>94.0% (sum of tautomers)

Description

3-Methyl-2,4-pentanedione involves in asymmetric substitution of 1,3-diphenyl-2-propenyl acetate catalyzed by amphiphilic resin-supported monodentate phosphine ligands can occur.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2,4-pentanedione involves in asymmetric substitution of 1,3-diphenyl-2-propenyl acetate catalyzed by amphiphilic resin-supported monodentate phosphine ligands can occur.

Solubility
Soluble in water.

Notes
Air & heat sensitive. Do not store together with strong bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 3
    Label Elements
    Signal Word

    Warning

    Physical Hazards

    H226 - Flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P403 + P233 - Store in a well-ventilated place. Keep container tightly closed