9-Acetylanthracene, 96%
9-Acetylanthracene, 96%
9-Acetylanthracene, 96%
Thermo Scientific Chemicals

9-Acetylanthracene, 96%

CAS: 784-04-3 | C16H12O | 220.271 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L01278.06
also known as L01278-06
Price (EUR)
46,60
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
46,60
Each
Chemical Identifiers
CAS784-04-3
IUPAC Name1-(anthracen-9-yl)ethan-1-one
Molecular FormulaC16H12O
InChI KeyNXXNVJDXUHMAHU-UHFFFAOYSA-N
SMILESCC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
Melting Point (clear melt)70-78?C
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream to yellow to brown
Identification (FTIR)Conforms
9-Acetylanthracene is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Acetylanthracene is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Insoluble in water.

Notes
Store at 15 - 30°C. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Kumao Hamanoue; Kazuo Nakajima; Toshiharu Hidaka; Toshihiro Nakayama and Hiroshi Teranishi. Time-dependent Picosecond Transient Absorption Spectra of 9-Acetylanthracene, Benzophenone and Acridine in Solution.Laser Chem.1984, 4 287-295.
  2. K Anderson; H Becker; LM Engelhardt; L Hansen and AH White. Molecular geometry and crystal structure of 9-acetylanthracene.Australian Journal of Chemistry.1984, 37 (6), 1337 - 1340.