2-Methyl-2-thiazoline, 97%
2-Methyl-2-thiazoline, 97%
2-Methyl-2-thiazoline, 97%
Thermo Scientific Chemicals

2-Methyl-2-thiazoline, 97%

CAS: 2346-00-1 | C4H7NS | 101.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L01329.14
also known as L01329-14
Price (EUR)
53,60
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
53,60
Each
Chemical Identifiers
CAS2346-00-1
IUPAC Name2-methyl-4,5-dihydro-1,3-thiazole
Molecular FormulaC4H7NS
InChI KeyJUIQOABNSLTJSW-UHFFFAOYSA-N
SMILESCC1=NCCS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless to yellow
Assay (GC)>96.0%
Refractive Index1.5160-1.5210
Formliquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only