7-Nitro-1-tetralone, 97%
7-Nitro-1-tetralone, 97%
7-Nitro-1-tetralone, 97%
Thermo Scientific Chemicals

7-Nitro-1-tetralone, 97%

CAS: 40353-34-2 | C10H9NO3 | 191.186 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number L01853.04
also known as L01853-04
Price (EUR)
80,00
Each
Quantity:
2 g
Request bulk or custom format
Price (EUR)
80,00
Each
Chemical Identifiers
CAS40353-34-2
IUPAC Name7-nitro-1,2,3,4-tetrahydronaphthalen-1-one
Molecular FormulaC10H9NO3
InChI KeyGWAQYWSNCVEJMW-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C2CCCC(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)White to cream to orange to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only