2-Methylcyclohexylamine, cis + trans, 97%
2-Methylcyclohexylamine, cis + trans, 97%
2-Methylcyclohexylamine, cis + trans, 97%
2-Methylcyclohexylamine, cis + trans, 97%
Thermo Scientific Chemicals

2-Methylcyclohexylamine, cis + trans, 97%

CAS: 7003-32-9 | C7H15N | 113.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L02285.09
also known as L02285-09
Price (EUR)
26,40
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (EUR)
26,40
Each
Add to cart
Chemical Identifiers
CAS7003-32-9
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
Refractive Index1.4550-1.4600 @ 20?C
2-Methylcyclohexylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.
RUO – Research Use Only

General References:

  1. Michael W. Rathke.; Naoto Inoue.; K. R. Varma.; Herbert C. Brown. A Stereospecific Synthesis of Alicyclic and Bicyclic Amines via Hydroboration. J. Am. Chem. Soc. 1966, 88 (12), 2870-2871.
  2. Rota F.; Prins R. Mechanism of the C-N-bond breaking in the hydrodenitrogenation of methylcyclohexylamine over sulfided NiMo/γ-Al2O3. J. Mol. Catal. A: Chem. 2000, 162(1), 367-74.