Dimethyl allylmalonate, 97%
Dimethyl allylmalonate, 97%
Dimethyl allylmalonate, 97%
Thermo Scientific Chemicals

Dimethyl allylmalonate, 97%

CAS: 40637-56-7 | C8H12O4 | 172.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L02376.09
also known as L02376-09
Price (EUR)
45,00
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
45,00
Each
Chemical Identifiers
CAS40637-56-7
IUPAC Name1,3-dimethyl 2-(prop-2-en-1-yl)propanedioate
Molecular FormulaC8H12O4
InChI KeyVZNFVLWVVHHMBG-UHFFFAOYSA-N
SMILESCOC(=O)C(CC=C)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4325-1.4375 @ 20?C
FormLiquid
Dimethyl allylmalonate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl allylmalonate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Notes
Protect from moisture, heat. Incomaptible with oxidizing agents. Store in cool place.
RUO – Research Use Only