5-Aminoindane, 97%
5-Aminoindane, 97%
5-Aminoindane, 97%
Thermo Scientific Chemicals

5-Aminoindane, 97%

CAS: 24425-40-9 | C9H11N | 133.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L02504.09
also known as L02504-09
Price (EUR)
75,60
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
75,60
Each
Chemical Identifiers
CAS24425-40-9
IUPAC Name2,3-dihydro-1H-inden-5-amine
Molecular FormulaC9H11N
InChI KeyLEWZOBYWGWKNCK-UHFFFAOYSA-N
SMILESNC1=CC=C2CCCC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown or pale pink
FormFused solid or clear liquid as melt
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only