2,6-Pyridinedimethanol, 98%
2,6-Pyridinedimethanol, 98%
2,6-Pyridinedimethanol, 98%
Thermo Scientific Chemicals

2,6-Pyridinedimethanol, 98%

CAS: 1195-59-1 | C7H9NO2 | 139.154 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L02527.06
also known as L02527-06
Price (EUR)
74,20
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
74,20
Each
Chemical Identifiers
CAS1195-59-1
IUPAC Name[6-(hydroxymethyl)pyridin-2-yl]methanol
Molecular FormulaC7H9NO2
InChI KeyWWFMINHWJYHXHF-UHFFFAOYSA-N
SMILESOCC1=CC=CC(CO)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)White to cream to yellow or pale brown
FormCrystalline powder or powder
Assay (GC)≥97.5%
Melting Point (clear melt)108-118?C
2,6-Pyridinedimethanol was used in the preparation of Fe(III) complexes [Fe(18)O(6)(OH)(8)(pdm)(10)(pdmH)(4)(H(2)O)(4)](ClO(4))(10) and Fe(9)O(4)(OH)(2)(O(2)CMe)(10)(pdm)(pdmH)(4)](NO(3)). It is a ligand used in the synthesis of a variety of metal complexes and catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Pyridinedimethanol was used in the preparation of Fe(III) complexes [Fe(18)O(6)(OH)(8)(pdm)(10)(pdmH)(4)(H(2)O)(4)](ClO(4))(10) and Fe(9)O(4)(OH)(2)(O(2)CMe)(10)(pdm)(pdmH)(4)](NO(3)). It is a ligand used in the synthesis of a variety of metal complexes and catalysts.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Taketo Taguchi, et al. Unusual Fe9 and Fe18 structural types from the use of 2,6-pyridinedimethanol in Fe(III) cluster chemistry.Dalton Trans.,2010,39(38), 9131-9139.
  2. Joel M. Hawkins, et al. Dioxomolybdenum(VI)-substituted 2,6-pyridinedimethanol complexes: new five-coordinate species.Inorg. Chem.,1986,25(9), 1501-1503.