N'-Ethyl-N,N-dimethylethylenediamine, 98%
N'-Ethyl-N,N-dimethylethylenediamine, 98%
N'-Ethyl-N,N-dimethylethylenediamine, 98%
Thermo Scientific Chemicals

N'-Ethyl-N,N-dimethylethylenediamine, 98%

CAS: 123-83-1 | C6H18N2 | 118.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L03180.14
also known as L03180-14
Price (EUR)
166,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
166,00
Each
Chemical Identifiers
CAS123-83-1
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4200-1.4240 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Reatant in the synthesis of tripod amphiphile. N'-Ethyl-N,N-dimethylethylenediamine is used as catalysts for olefin aziridination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of tripod amphiphile. N′-Ethyl-N,N-dimethylethylenediamine is used as catalysts for olefin aziridination.

Notes
Air Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide.
RUO – Research Use Only

General References:

  1. Diaz C, et al. Reactions Between Dinuclear CuII-NiII Complexes and the Nitrito Ligand: A New One-Dimensional Complex [Cu(oxpn)Ni(μ-NO2)(trimen)]n(ClO4)n with Nitrite Acting as a Tridentate Bridging Ligand and Ferro- and Antiferromagnetic Alternation. Eur. J. Inorg. Chem. 2000, 2000 (4), 675-681.
  2. Thirumanasekaran Dhanalakshmi,; Eringathodi Suresh,; Mallayan Palaniandav.Olefin aziridination by copper(II) complexes: Effect of redox potential on catalytic activity. Inorganica Chimica Acta. 2011, 365(1), 143-151.