2',4',6'-Trihydroxyacetophenone hydrate, 98+%
2',4',6'-Trihydroxyacetophenone hydrate, 98+%
2',4',6'-Trihydroxyacetophenone hydrate, 98+%
Thermo Scientific Chemicals

2',4',6'-Trihydroxyacetophenone hydrate, 98+%

CAS: 480-66-0 | C8H8O4 | 168.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L04156.14
also known as L04156-14
Price (EUR)
157,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
157,00
Each
Specifications
Beilstein1911197
CAS480-66-0
ChEBICHEBI:64344
Chemical Name or Material2',4',6'-Trihydroxyacetophenone hydrate
EINECS Number207-556-5
View more
2',4',6'-Trihydroxyacetophenone hydrate is a compound that enhances bile secretion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,4′,6′-Trihydroxyacetophenone hydrate is a compound that enhances bile secretion.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
Hans R. Kricheldorf.; Michael Rabenstein.; M. Maskos.; M. Schmidt. Macrocycles. 15. The Role of Cyclization in Kinetically Controlled Polycondensations. 1. Polyester Syntheses. Macromolecules. 2001, 34 (4), 713-722.