Chloro(methyl)diphenylsilane, 97%
Chloro(methyl)diphenylsilane, 97%
Chloro(methyl)diphenylsilane, 97%
Thermo Scientific Chemicals

Chloro(methyl)diphenylsilane, 97%

CAS: 144-79-6 | C13H13ClSi | 232.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L04162.18
also known as L04162-18
Price (EUR)
106,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
106,00
Each
Chemical Identifiers
CAS144-79-6
IUPAC Namechloro(methyl)diphenylsilane
Molecular FormulaC13H13ClSi
InChI KeyOJZNZOXALZKPEA-UHFFFAOYSA-N
SMILESC[Si](Cl)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Refractive Index1.5710-1.5760 @ 20?C
FormLiquid
Chloro(methyl)diphenylsilane is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloro(methyl)diphenylsilane is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from strong bases. Incompatible with moisture and bases.
RUO – Research Use Only

General References:

  1. Filippo Marchioni; Kurt Star; Erik Menke; Thierry Buffeteau; Laurent Servant; Bruce Dunn and Fred Wud. Protection of Lithium Metal Surfaces Using Chlorosilanes.Anal. Chem.2007, 23 (23), 11597-11602.
  2. Silylating agent (see Appendix 4) for formation, in the presence of a base such as imidazole in DMF, of DPMS ethers, intermediate in hydrolytic stabilty between TMS and TBDMS. DPMS ethers are incompatible with acid, base, BuLi, Cr(VI) oxidants and LIAlH4, but stable to silica chromatography: J. Org. Chem., 52, 165 (1987). They can be cleaved with mild acid or base, or F-. Selective cleavage in high yield in the presence of TBDMS or TBDPS ethers with NaN3 in DMF was reported: J. Chem. Soc., Chem. Commun., 381 (1989).
  3. In reactions with lithiated esters and lactones C-silylation predominates. The resulting ɑ-DPMS esters behave as vinyl dication equivalents: J. Am. Chem. Soc., 103, 2418 (1981); Pure Appl. Chem., 62, 2021 (1990), useful precursors of, e.g.: terminal alkenes: Synth. Commun., 13, 1163 (1983); 1,1-disubstituted alkenes: Tetrahedron Lett., 23, 271 (1982); Org. Synth. Coll., 8, 474 (1993); ß-keto silanes and ketones: J. Org. Chem., 50, 5260 (1985); 58, 2718 (1993):
  4. TBDMS esters react with aldehydes and ketones to give ß-hydroxy silanes, which undergo Peterson elimination to ɑ-substituted ɑß-unsaturated esters: J. Org. Chem., 49, 3385 (1984).