Polygalacturonic acid, M.W. 25,000-50,000
Polygalacturonic acid, M.W. 25,000-50,000
Polygalacturonic acid, M.W. 25,000-50,000
Thermo Scientific Chemicals

Polygalacturonic acid, M.W. 25,000-50,000

CAS: 25990-10-7 | (C6H8O6)n | 194.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number L04206.22
also known as L04206-22
Price (EUR)
247,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
247,00
Each
Chemical Identifiers
CAS25990-10-7
Molecular Formula(C6H8O6)n
SMILESO[C@@H]1[C@@H](O)[C@@H](O-*)[C@H](OC1-*)C(O)=O
MDL NumberMFCD00131972
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (unspecified)≥86.0% (dry wt. basis)
Loss on Drying≤10.0% (105?C/constant wt.)
FormPowder
Identification (FTIR)Conforms
View more
Polygalacturonic acid is reagent which sequesters metal ions and produces gels that can be used for the production of biomolecules. It is used for drug release or to immobilize cells and enzymes . Polygalacturonic acid may be used to identify, differentiate and characterized plant cell wall degrading polygalacturonase(s) from fungi.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only