n-Butyl phenyl ether, 99%
n-Butyl phenyl ether, 99%
n-Butyl phenyl ether, 99%
Thermo Scientific Chemicals

n-Butyl phenyl ether, 99%

CAS: 1126-79-0 | C10H14O | 150.221 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L04703.14
also known as L04703-14
Price (EUR)
49,70
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
49,70
Each
Chemical Identifiers
CAS1126-79-0
IUPAC Namebutoxybenzene
Molecular FormulaC10H14O
InChI KeyYFNONBGXNFCTMM-UHFFFAOYSA-N
SMILESCCCCOC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)>98.5%
Refractive Index1.4950-1.4990 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only