(-)-Shikimic acid, 98%
(-)-Shikimic acid, 98%
(-)-Shikimic acid, 98%
Thermo Scientific Chemicals

(-)-Shikimic acid, 98%

CAS: 138-59-0 | C7H9O5- | 173.144 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L04848.MD
also known as L04848-MD
Price (EUR)
35,70
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
35,70
Each
Chemical Identifiers
CAS138-59-0
IUPAC Name3,4,5-trihydroxycyclohex-1-ene-1-carboxylic acid
Molecular FormulaC7H10O5
InChI KeyJXOHGGNKMLTUBP-UHFFFAOYNA-N
SMILESOC1CC(=CC(O)C1O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Water Content (Karl Fischer Titration)≤1.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only