1-(3-Aminopropyl)-4-methylpiperazine, 98%
1-(3-Aminopropyl)-4-methylpiperazine, 98%
1-(3-Aminopropyl)-4-methylpiperazine, 98%
1-(3-Aminopropyl)-4-methylpiperazine, 98%
Thermo Scientific Chemicals

1-(3-Aminopropyl)-4-methylpiperazine, 98%

CAS: 4572-03-6 | C8H19N3 | 157.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L04876.06
also known as L04876-06
Price (EUR)
241,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
241,00
Each
Add to cart
Chemical Identifiers
CAS4572-03-6
IUPAC Name3-(4-methylpiperazin-1-yl)propan-1-amine
Molecular FormulaC8H19N3
InChI KeyRGUABPVONIGVAT-UHFFFAOYSA-N
SMILESCN1CCN(CCCN)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)>97.5%
Refractive Index1.4800-1.4840 @ 20?C
1-(3-Aminopropyl)-4-methylpiperazine is used as a precursor in organic synthesis for the preparation of furan-2-carboxylic acid [3-(4-methyl-piperazin-1-yl)-propyl]-amide by reacting with furan-2-carbonyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Aminopropyl)-4-methylpiperazine is used as a precursor in organic synthesis for the preparation of furan-2-carboxylic acid [3-(4-methyl-piperazin-1-yl)-propyl]-amide by reacting with furan-2-carbonyl chloride.

Notes
Air sensitive. Keep in a dry, cool and well-ventilated place. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Meyer, R. A.; Meyer, R. S. M.; Green, J. J. An automated multidimensional thin film stretching device for the generation of anisotropic polymeric micro- and nanoparticles. J. Biomed. Mater. Res. Part A 2015, 103 (8), 2747-2757.
  2. Bishop, C. J.; Tzeng, S. Y.; Green, J. J. Degradable polymer-coated gold nanoparticles for co-delivery of DNA and siRNA. Acta Biomater. 2015, 11, 393-403.