Molecular sieves, 3A, 2-5 mm (0.08-0.20in) beads
Molecular sieves, 3A, 2-5 mm (0.08-0.20in) beads
Molecular sieves, 3A, 2-5 mm (0.08-0.20in) beads
Thermo Scientific Chemicals

Molecular sieves, 3A, 2-5 mm (0.08-0.20in) beads

CAS: 308080-99-1 | AlNaO6Si2 | 202.135 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number L05359.0B
also known as L05359-0B
Price (EUR)
127,65
キャンペーン価格
142,00
Save 14,35 (10%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (EUR)
127,65
キャンペーン価格
142,00
Save 14,35 (10%)
Each
Chemical Identifiers
CAS308080-99-1
IUPAC NameMolecular sieves 3A
MDL NumberMFCD00131613
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale grey to cream to brown
FormBeads
Residual water≤1.5%
Particle Size2-5mm
Water adsorption≥20.0% (60% relative humidity)
Molecular sieves, 3A is used as a desiccant in petroleum and chemical industries. It is useful for the drying of unsaturated hydrocarbons such as ethylene, propylene and butadiene. It is also used in the removal of water from ethanol. It finds application in the drying of certain chemicals namely ethanol, refrigerants and natural gas. It plays an important role in selective adsorption of water in insulated glass (IG) and polyurethane. It has been considered as a general-purpose drying agent in polar and non polar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only