(R)-(+)-1-Phenyl-1-propanol, 99%
(R)-(+)-1-Phenyl-1-propanol, 99%
(R)-(+)-1-Phenyl-1-propanol, 99%
Thermo Scientific Chemicals

(R)-(+)-1-Phenyl-1-propanol, 99%

CAS: 1565-74-8 | C9H12O | 136.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number L05681.ME
also known as L05681-ME
Price (EUR)
106,00
Each
Quantity:
500 mg
Request bulk or custom format
Price (EUR)
106,00
Each
Chemical Identifiers
CAS1565-74-8
IUPAC Name1-phenylpropan-1-ol
Molecular FormulaC9H12O
InChI KeyDYUQAZSOFZSPHD-UHFFFAOYNA-N
SMILESCCC(O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5185-1.5215 @ 20?C
Optical Rotation28.5? ± 0.5? (neat)
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
FormLiquid
For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only