Thermo Scientific Chemicals

Perfluoro(methyldecahydronaphthalene), tech. 85%, mixture of isomers, Thermo Scientific Chemicals

Catalog number: L05862.14
25 g, Each
Thermo Scientific Chemicals

Perfluoro(methyldecahydronaphthalene), tech. 85%, mixture of isomers, Thermo Scientific Chemicals

Catalog number: L05862.14
25 g, Each
Quantity
Catalog number: L05862.14
also known as L05862-14
Price (EUR)

Chemical Identifiers

CAS
51294-16-7
IUPAC Name
1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8a-heptadecafluoro-8-(trifluoromethyl)-decahydronaphthalene
Molecular Formula
C11F20
InChI Key
LWRNQOBXRHWPGE-UHFFFAOYNA-N
SMILES
FC(F)(F)C1(F)C(F)(F)C(F)(F)C(F)(F)C2(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C12F
Form
Liquid
Refractive Index
1.3140-1.3220 @ 20?C
Appearance (Color)
Clear, colorless
Assay (GC)
>83.0% (mixture of isomers)
Comment
Mixture of 1- and 2- isomers, remainder other fluorocarbons.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text