Methyl 3,4,5-trihydroxybenzoate, 98%
Methyl 3,4,5-trihydroxybenzoate, 98%
Methyl 3,4,5-trihydroxybenzoate, 98%
Thermo Scientific Chemicals

Methyl 3,4,5-trihydroxybenzoate, 98%

CAS: 99-24-1 | C8H8O5 | 184.147 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number L05926.30
also known as L05926-30
Price (EUR)
146,00
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
146,00
Each
Chemical Identifiers
CAS99-24-1
IUPAC Namemethyl 3,4,5-trihydroxybenzoate
Molecular FormulaC8H8O5
InChI KeyFBSFWRHWHYMIOG-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(O)=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥97.5%
Appearance (Color)White to cream or pale pink
FormCrystals or powder or crystalline powder
Methyl 3,4,5-trihydroxybenzoate exhibits potent antitumor activities by inhibiting tumor infiltration of CD4+CD25+ regulatory T cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3,4,5-trihydroxybenzoate exhibits potent antitumor activities by inhibiting tumor infiltration of CD4+CD25+ regulatory T cells.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Pitchaon Maisuthisakul; Michael H Gordon. Characterization and storage stability of the extract of Thai mango (Mangifera indica Linn. Cultivar Chok-Anan) seed kernels. Journal of Food Science and Technology.2014, 51, (8), 1453-1462.
  2. Lee H; Lee H; Kwon Y; Lee JH; Kim J; Shin MK; Kim SH; Bae H. Methyl gallate exhibits potent antitumor activities by inhibiting tumor infiltration of CD4+CD25+ regulatory T cells. J Immunol.2010, 185, (11), 6698-6705.