2-Isopropoxyphenol, 97%
2-Isopropoxyphenol, 97%
2-Isopropoxyphenol, 97%
Thermo Scientific Chemicals

2-Isopropoxyphenol, 97%

CAS: 4812-20-8 | C9H12O2 | 152.193 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L07360.06
also known as L07360-06
Price (EUR)
41,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
41,00
Each
Chemical Identifiers
CAS4812-20-8
IUPAC Name2-(propan-2-yloxy)phenol
Molecular FormulaC9H12O2
InChI KeyZNCUUYCDKVNVJH-UHFFFAOYSA-N
SMILESCC(C)OC1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.5125-1.5175 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to pale yellow to yellow to orange to red or pale pink
2-Isopropoxyphenol is a metabolite of propoxur. It was used as a target chemical in developing a method to measure the urinary phenolic metabolites

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Isopropoxyphenol is a metabolite of propoxur. It was used as a target chemical in developing a method to measure the urinary phenolic metabolites

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Hans Weiss. Flow-injection spectrophometric determination of propoxur with p-aminophenol. Analytica Chimica Acta. 1957, 49, (7), 1089-1090.
  2. Karim D. Khalaf.; J. Sancenón.; M. de la Guardia. Zinc hydroxystannate as an alternative synergist to antimony trioxide in polyester resins containing halogenated flame retardants. Analytica Chimica Acta. 1992, 266, (1), 119-126.