1,2-Epoxyoctadecane, tech. 85%
1,2-Epoxyoctadecane, tech. 85%
1,2-Epoxyoctadecane, tech. 85%
Thermo Scientific Chemicals

1,2-Epoxyoctadecane, tech. 85%

CAS: 7390-81-0 | C18H36O | 268.485 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L07622.22
also known as L07622-22
Price (EUR)
115,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
115,00
Each
Chemical Identifiers
CAS7390-81-0
IUPAC Name2-hexadecyloxirane
Molecular FormulaC18H36O
InChI KeyQBJWYMFTMJFGOL-UHFFFAOYNA-N
SMILESCCCCCCCCCCCCCCCCC1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or fused solid or lumps
Assay (GC)≥80.0%
1,2-Epoxyoctadecane can be used to produce octadecane-1,2-diol at heating. It is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxyoctadecane can be used to produce octadecane-1,2-diol at heating. It is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Insoluble in water.

Notes
Protect from moisture. Store away from incompatible materials such as oxidizing agents. Moisture-sensitive. Incompatible with strong acids, peroxides, caustics.
RUO – Research Use Only