1-(n-Butyl)imidazole, 99%
1-(n-Butyl)imidazole, 99%
1-(n-Butyl)imidazole, 99%
Thermo Scientific Chemicals

1-(n-Butyl)imidazole, 99%

CAS: 4316-42-1 | C7H12N2 | 124.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L07793.22
also known as L07793-22
Price (EUR)
106,65
Online exclusive
118,00
Save 11,35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
106,65
Online exclusive
118,00
Save 11,35 (10%)
Each
Chemical Identifiers
CAS4316-42-1
IUPAC Name1-butyl-1H-imidazole
Molecular FormulaC7H12N2
InChI KeyMCMFEZDRQOJKMN-UHFFFAOYSA-N
SMILESCCCCN1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4785-1.4815 @ 20?C
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to yellow
1-(n-Butyl)imidazole is used in the synthesis of a heterocyclic mesomeric betaine and 1-(1-butyl-3-imidazolio)propane-3-sulfonate (BIm3S). It acts as an N-coordinated ligand. It is used in the chiral separation of amino acids and anionic pharmaceuticals by capillary electrophoresis. It is used for preparation of substituted phenyl-containing imidazolium ionic liquid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(n-Butyl)imidazole is used in the synthesis of a heterocyclic mesomeric betaine and 1-(1-butyl-3-imidazolio)propane-3-sulfonate (BIm3S). It acts as an N-coordinated ligand. It is used in the chiral separation of amino acids and anionic pharmaceuticals by capillary electrophoresis. It is used for preparation of substituted phenyl-containing imidazolium ionic liquid.

Solubility
Soluble in Chloroform, Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

General References:

  1. Nina Gonsior; Fabian Mohr; Helmut Ritter. Synthesis of mesomeric betaine compounds with imidazolium-enolate structure.Beilstein Journal of Organic Chemistry.2012, 8 390-397.
  2. G M Smith; G G Duncan. A study of intravascular platelet aggregation by continuous platelet counting.Thrombosis Research.1981, 23 (3), 275-288.
  3. Preferred ligand in a Cu(I)-catalyzed procedure for cyanation of heteroaryl bromides with potassium hexacyanoferrate(II), as an alternative to conventional routes with highly toxic metal cyanides: Synlett, 555 (2007).