N-Phenyliminodiacetic acid, 97%
N-Phenyliminodiacetic acid, 97%
N-Phenyliminodiacetic acid, 97%
Thermo Scientific Chemicals

N-Phenyliminodiacetic acid, 97%

CAS: 1137-73-1 | C10H11NO4 | 209.201 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L08437.14
also known as L08437-14
Price (EUR)
246,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
246,00
Each
Chemical Identifiers
CAS1137-73-1
IUPAC Name2-[(carboxymethyl)(phenyl)amino]acetic acid
Molecular FormulaC10H11NO4
InChI KeyGQBWTAGIANQVGB-UHFFFAOYSA-N
SMILESOC(=O)CN(CC(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or granules
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White to cream to yellow to pale green or pale purple to pale pink to gray
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only