5,5-Dimethyl-1,3-dioxan-2-one, tech.
5,5-Dimethyl-1,3-dioxan-2-one, tech.
5,5-Dimethyl-1,3-dioxan-2-one, tech.
5,5-Dimethyl-1,3-dioxan-2-one, tech.
Thermo Scientific Chemicals

5,5-Dimethyl-1,3-dioxan-2-one, tech.

CAS: 3592-12-9 | C6H10O3 | 130.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L08767.03
also known as L08767-03
Price (EUR)
43,50
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
43,50
Each
Add to cart
Chemical Identifiers
CAS3592-12-9
IUPAC Name5,5-dimethyl-1,3-dioxan-2-one
Molecular FormulaC6H10O3
InChI KeyJRFXQKZEGILCCO-UHFFFAOYSA-N
SMILESCC1(C)COC(=O)OC1
View more
SpecificationsSpecification SheetSpecification Sheet
Purity (DSC)≥83.0%
Appearance (Color)White
CommentContains varying amounts of oligomers
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only