(+/-)-1-(1-Naphthyl)ethylamine, 98%
(+/-)-1-(1-Naphthyl)ethylamine, 98%
(+/-)-1-(1-Naphthyl)ethylamine, 98%
Thermo Scientific Chemicals

(+/-)-1-(1-Naphthyl)ethylamine, 98%

CAS: 42882-31-5 | C12H13N | 171.243 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L09342.14
also known as L09342-14
Price (EUR)
166,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
166,00
Each
Chemical Identifiers
CAS42882-31-5
IUPAC Name1-(naphthalen-1-yl)ethan-1-amine
Molecular FormulaC12H13N
InChI KeyRTCUCQWIICFPOD-UHFFFAOYNA-N
SMILESCC(N)C1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)Clear colorless to yellow
Refractive Index1.6205-1.6255 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only