2,3-Dichlorobenzoyl chloride, 98%
2,3-Dichlorobenzoyl chloride, 98%
2,3-Dichlorobenzoyl chloride, 98%
Thermo Scientific Chemicals

2,3-Dichlorobenzoyl chloride, 98%

CAS: 2905-60-4 | C7H3Cl3O | 209.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L09506.22
also known as L09506-22
Price (EUR)
385,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
385,00
Each
Chemical Identifiers
CAS2905-60-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to pale brown
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)27.0-34.0?C
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder or fused solid or granules or clear liquid as melt
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only