1,4-Bis[(trimethylsilyl)ethynyl]benzene, 98%
1,4-Bis[(trimethylsilyl)ethynyl]benzene, 98%
1,4-Bis[(trimethylsilyl)ethynyl]benzene, 98%
Thermo Scientific Chemicals

1,4-Bis[(trimethylsilyl)ethynyl]benzene, 98%

CAS: 17938-13-5 | C16H22Si2 | 270.522 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09709.03
also known as L09709-03
Price (EUR)
32,70
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
32,70
Each
Chemical Identifiers
CAS17938-13-5
IUPAC Nametrimethyl(2-{4-[2-(trimethylsilyl)ethynyl]phenyl}ethynyl)silane
Molecular FormulaC16H22Si2
InChI KeyCMTMWEXUJQSPCA-UHFFFAOYSA-N
SMILESC[Si](C)(C)C#CC1=CC=C(C=C1)C#C[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to yellow to orange to brown
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)118.0-124.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only