2,4-Dimethoxyphenyl isocyanate, 97%
2,4-Dimethoxyphenyl isocyanate, 97%
2,4-Dimethoxyphenyl isocyanate, 97%
Thermo Scientific Chemicals

2,4-Dimethoxyphenyl isocyanate, 97%

CAS: 84370-87-6 | C9H9NO3 | 179.175 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09762.03
also known as L09762-03
Price (EUR)
30,20
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
30,20
Each
Chemical Identifiers
CAS84370-87-6
IUPAC Name1-isocyanato-2,4-dimethoxybenzene
Molecular FormulaC9H9NO3
InChI KeyWRAZHLRMDRTLOZ-UHFFFAOYSA-N
SMILESCOC1=CC=C(N=C=O)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps or fused solid or clear liquid as melt
Melting Point (clear melt)26.0-35.0?C
Appearance (Color)White to pale cream to pale yellow
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only