Zinc-copper couple, Copper content typically ca 1-4%
Zinc-copper couple, Copper content typically ca 1-4%
Zinc-copper couple, Copper content typically ca 1-4%
Thermo Scientific Chemicals

Zinc-copper couple, Copper content typically ca 1-4%

CAS: 53801-63-1 | CuZn | 128.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L09811.14
also known as L09811-14
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS53801-63-1
SpecificationsSpecification SheetSpecification Sheet
Elemental Analysis(Copper, Cu) : ≥0.5 to ≤4.0%
Appearance (Color)Grey to dark grey
Elemental Analysis(Zinc, Zn) : ≥90.0%
FormPowder
Zinc-copper couple is used in the Simmons-Smith cyclopropanation reaction, Reformatsky reaction. It is used as a chemical for the selective, stereospecific reduction of alkynes to cis-alkenes. It is used in for deiodination of iodotributylstannylalkanes. In the presence of a free-radical initiator, it promotes the formylation of perfluoroalkyl iodides with DMF to give fluorinated aldehydes in high yield. It is used in dermatology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc-copper couple is used in the Simmons-Smith cyclopropanation reaction, Reformatsky reaction. It is used as a chemical for the selective, stereospecific reduction of alkynes to cis-alkenes. It is used in for deiodination of iodotributylstannylalkanes. In the presence of a free-radical initiator, it promotes the formylation of perfluoroalkyl iodides with DMF to give fluorinated aldehydes in high yield. It is used in dermatology.

Solubility
Insoluble in water.

Notes
Store at 4°C. Moisture Sensitive. Do not store together with acids. Protect from humidity and water. Incompatible materials are amines, cadmium, chlorinated solvents, Sulfur compounds.
RUO – Research Use Only

General References:

  1. Eugene LeGoff. Cyclopropanes from an Easily Prepared, Highly Active Zinc—Copper Couple, Dibromomethane, and Olefins.J. Org. Chem.1964, 29 (7), 2048-2050.
  2. S. Morris Kupchan; Masao Maruyama. Reductive elimination of epoxides to olefins with zinc-copper couple.J. Org. Chem.1971, 36 (9), 1187-1191.
  3. For use in the Simmons-Smith cyclopropanation reaction, see Diiodomethane, A15457. For use in the Reformatsky reaction, see: Synthesis, 698 (1977).
  4. Used for the selective, stereospecific reduction of alkynes to cis-alkenes: Chem. Ind. (London), 143 (1957). For deiodination of iodotributylstannylalkanes, see: Synlett., 891 (1992).
  5. In the presence of a free-radical initiator, promotes the formylation of perfluoroalkyl iodides with DMF to give fluorinated aldehydes in high yield: J. Fluorine Chem., 63, 217 (1993).