Methyl 2-naphthoate, 99%
Methyl 2-naphthoate, 99%
Methyl 2-naphthoate, 99%
Methyl 2-naphthoate, 99%
Thermo Scientific Chemicals

Methyl 2-naphthoate, 99%

CAS: 2459-25-8 | C12H10O2 | 186.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09973.06
also known as L09973-06
Price (EUR)
67,49
Online Exclusive
79,40
Save 11,91 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
67,49
Online Exclusive
79,40
Save 11,91 (15%)
Each
Add to cart
Chemical Identifiers
CAS2459-25-8
IUPAC Namemethyl naphthalene-2-carboxylate
Molecular FormulaC12H10O2
InChI KeyIODOXLXFXNATGI-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)74.0-80.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only