Methyl 2,2-dimethylacetoacetate, 99%
Methyl 2,2-dimethylacetoacetate, 99%
Methyl 2,2-dimethylacetoacetate, 99%
Thermo Scientific Chemicals

Methyl 2,2-dimethylacetoacetate, 99%

CAS: 38923-57-8 | C7H12O3 | 144.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L10917.06
also known as L10917-06
Price (EUR)
135,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
135,00
Each
Chemical Identifiers
CAS38923-57-8
IUPAC Namemethyl 2,2-dimethyl-3-oxobutanoate
Molecular FormulaC7H12O3
InChI KeyLVSDLZIEHYYLTC-UHFFFAOYSA-N
SMILESCOC(=O)C(C)(C)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4190-1.4220 @ 20?C
Assay (GC)≥98.5%
FormLiquid
Pyrimidine was synthesis from methyl 2, 2-dimethylacetoacetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrimidine was synthesis from methyl 2, 2-dimethylacetoacetate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
James, Stephen R.,; Colin B. Reese. Pyrimidine synthesis from methyl 2, 2-dimethylacetoacetate.. Tetrahedron Letters. 1975, 16(33), 2915-2916..