(Bromomethyl)cyclohexane, 98%
(Bromomethyl)cyclohexane, 98%
(Bromomethyl)cyclohexane, 98%
Thermo Scientific Chemicals

(Bromomethyl)cyclohexane, 98%

CAS: 2550-36-9 | C7H13Br | 177.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L11343.14
also known as L11343-14
Price (EUR)
89,20
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
89,20
Each
Chemical Identifiers
CAS2550-36-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Identification (FTIR)Conforms
CommentMaterial sourced in the U.S. and in other countries
FormLiquid
View more
(Bromomethyl)cyclohexane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Bromomethyl)cyclohexane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Philip Boudjouk.; Ratnasabapathy Sooriyakumaran.; Craig A. Kapfer. The reaction of magnesium with cis-1,3,5-tris(bromomethyl)cyclohexane. Evidence for a soluble tri-Grignard. Journal of Organometallic Chemistry. 1985, 281 (2-3), 21-23.
  2. George P Luke.; Dennis A Holt. Synthesis of (S)-5-(1-aminoethyl)-2-(cyclohexylmethoxy)benzamide. Tetrahedron: Asymmetry. 1999, 10 (22), 4393-4403.