n-Hexyl methacrylate, 97%, stab. with 100ppm 4-methoxyphenol
n-Hexyl methacrylate, 97%, stab. with 100ppm 4-methoxyphenol
n-Hexyl methacrylate, 97%, stab. with 100ppm 4-methoxyphenol
Thermo Scientific Chemicals

n-Hexyl methacrylate, 97%, stab. with 100ppm 4-methoxyphenol

CAS: 142-09-6 | C10H18O2 | 170.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L11569.22
also known as L11569-22
Price (EUR)
111,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
111,00
Each
Chemical Identifiers
CAS142-09-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4285-1.4335 @20?C
Solution Test10% in methanol: clear
CommentStabilized with 50-150ppm 4-methoxyphenol (MEHQ)
FormLiquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only