5-Chloro-2,4-dinitrotoluene, 97%
5-Chloro-2,4-dinitrotoluene, 97%
5-Chloro-2,4-dinitrotoluene, 97%
5-Chloro-2,4-dinitrotoluene, 97%
Thermo Scientific Chemicals

5-Chloro-2,4-dinitrotoluene, 97%

CAS: 51676-74-5 | C7H5ClN2O4 | 216.577 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L11808.06
also known as L11808-06
Price (EUR)
64,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
64,00
Each
Add to cart
Chemical Identifiers
CAS51676-74-5
IUPAC Name1-chloro-5-methyl-2,4-dinitrobenzene
Molecular FormulaC7H5ClN2O4
InChI KeyKPDPGZNHKMJEFZ-UHFFFAOYSA-N
SMILESCC1=CC(Cl)=C(C=C1[N+]([O-])=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
Melting Point (clear melt)83.5-92.5°C
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only