Diacetin, mixed isomers, tech. ca 50%, remainder triacetin and monoacetin
Diacetin, mixed isomers, tech. ca 50%, remainder triacetin and monoacetin
Diacetin, mixed isomers, tech. ca 50%, remainder triacetin and monoacetin
Thermo Scientific Chemicals

Diacetin, mixed isomers, tech. ca 50%, remainder triacetin and monoacetin

CAS: 25395-31-7 | C7H12O5
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number L12708.30
also known as L12708-30
Price (EUR)
38,20
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
38,20
Each
Chemical Identifiers
CAS25395-31-7
IUPAC Name1-(acetyloxy)-3-hydroxypropan-2-yl acetate
Molecular FormulaC7H12O5
InChI KeyUXDDRFCJKNROTO-UHFFFAOYNA-N
SMILESCC(=O)OCC(CO)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>45.0% (main component)
Appearance (Color)Clear, colorless
FormLiquid
Refractive Index1.4380-1.4480 @ 20°C
Diacetin is widely used as plasticizer. It has been used to design and evaluate gliclazide push-pull osmotic pump (PPOP) coated with aqueous colloidal polymer dispersions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diacetin is widely used as plasticizer. It has been used to design and evaluate gliclazide push-pull osmotic pump (PPOP) coated with aqueous colloidal polymer dispersions.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Xin Tang; Ling-yu Tai; Xing-gang Yang; Fen Chen; He-ming Xu; Wei-san Pan. In vitro and in vivo evaluation of gliclazide push-pull osmotic pump coated with aqueous colloidal polymer dispersions. Drug Development and Industrial Pharmacy. 2013, 39,(1), 67-76.
  2. Karel Hernandez; Eduardo Garcia-Verdugo; Raul Porcar; Roberto Fernandez-Lafuente. Hydrolysis of triacetin catalyzed by immobilized lipases: Effect of the immobilization protocol and experimental conditions on diacetin yield. Journal name. 2011, 48,(6-7), 510-517.