Thiophene-3-carbonitrile, 98%
Thiophene-3-carbonitrile, 98%
Thiophene-3-carbonitrile, 98%
Thiophene-3-carbonitrile, 98%
Thermo Scientific Chemicals

Thiophene-3-carbonitrile, 98%

CAS: 1641-09-4 | C5H3NS | 109.146 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L13115.14
also known as L13115-14
Price (EUR)
183,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
183,00
Each
Add to cart
Chemical Identifiers
CAS1641-09-4
IUPAC Namethiophene-3-carbonitrile
Molecular FormulaC5H3NS
InChI KeyGSXCEVHRIVLFJV-UHFFFAOYSA-N
SMILESN#CC1=CSC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5% (UK-sourced material)
CommentMaterial sourced in UK and US
Refractive Index1.5585-1.5635 @ 20?C (UK-sourced material)
Infrared spectrumConforms (US-sourced material)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only