2-Amino-4,4,4-trifluorobutyric acid, 97%
2-Amino-4,4,4-trifluorobutyric acid, 97%
2-Amino-4,4,4-trifluorobutyric acid, 97%
Thermo Scientific Chemicals

2-Amino-4,4,4-trifluorobutyric acid, 97%

CAS: 15959-93-0 | C4H6F3NO2 | 157.092 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number L13131.ME
also known as L13131-ME
Price (EUR)
125,00
Each
Quantity:
500 mg
Request bulk or custom format
Price (EUR)
125,00
Each
Chemical Identifiers
CAS15959-93-0
IUPAC Name2-amino-4,4,4-trifluorobutanoic acid
Molecular FormulaC4H6F3NO2
InChI KeyAQPCXCOPDSEKQT-UHFFFAOYNA-N
SMILESNC(CC(F)(F)F)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only