Thermo Scientific Chemicals

Di-p-toluoyl-D-tartaric acid, 98%, Thermo Scientific Chemicals

Catalog number: L13223.14
25 g, Each
Thermo Scientific Chemicals

Di-p-toluoyl-D-tartaric acid, 98%, Thermo Scientific Chemicals

Catalog number: L13223.14
25 g, Each
Quantity
Catalog number: L13223.14
also known as L13223-14
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
32634-68-7
IUPAC Name
(2S,3S)-2,3-bis(4-methylbenzoyloxy)butanedioic acid
Molecular Formula
C20H18O8
InChI Key
CMIBUZBMZCBCAT-HOTGVXAUSA-N
SMILES
CC1=CC=C(C=C1)C(=O)O[C@@H]([C@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O
Form
Crystals or powder or crystalline powder
Comment
Specification differs for U.S. and non-U.S. material where indicated
Optical Rotation
139.4 ± 0.1° (C=1 in ethanol, non-U.S. specification)
Water Content (Karl Fischer Titration)
≤3%
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text