2,2',5,5'-Tetramethylbiphenyl, 98%
2,2',5,5'-Tetramethylbiphenyl, 98%
2,2',5,5'-Tetramethylbiphenyl, 98%
2,2',5,5'-Tetramethylbiphenyl, 98%
Thermo Scientific Chemicals

2,2',5,5'-Tetramethylbiphenyl, 98%

CAS: 3075-84-1 | C16H18 | 210.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L13330.06
also known as L13330-06
Price (EUR)
238,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
238,00
Each
Add to cart
Specifications
Beilstein2046200
CAS3075-84-1
Chemical Name or Material2,2',5,5'-Tetramethylbiphenyl
EINECS Number000-000-0
Formula Weight210.32
View more
2,2',5,5'-Tetramethylbiphenyl is used to make other chemicals and as an intermediate in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′,5,5′-Tetramethylbiphenyl is used to make other chemicals and as an intermediate in pharmaceuticals.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. C. S. Chao.; C. H. Cheng.; C. T. Chang. New method for the preparation of activated nickel and cobalt powders and their application in biaryl synthesis. J. Org. Chem. 1983, 48 (25), 4904-4907.
  2. Kang C. Park.; Lawrence R. Dodd.; Kalle Levon.; T. K. Kwei. Conformational Study of Poly(p-phenylene) by Molecular Mechanics Minimization. Macromolecules. 1996, 29 (22), 149-7154.