2-(Methacryloyloxy)ethyltrimethylammonium chloride, 72% aq. soln., stab. with 150-200 ppm 4-methoxyphenol
2-(Methacryloyloxy)ethyltrimethylammonium chloride, 72% aq. soln., stab. with 150-200 ppm 4-methoxyphenol
2-(Methacryloyloxy)ethyltrimethylammonium chloride, 72% aq. soln., stab. with 150-200 ppm 4-methoxyphenol
Thermo Scientific Chemicals

2-(Methacryloyloxy)ethyltrimethylammonium chloride, 72% aq. soln., stab. with 150-200 ppm 4-methoxyphenol

CAS: 5039-78-1 | C9H18ClNO2 | 207.698 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number L13523.22
also known as L13523-22
Price (EUR)
32,70
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
32,70
Each
Chemical Identifiers
CAS5039-78-1
IUPAC Nametrimethyl({2-[(2-methylprop-2-enoyl)oxy]ethyl})azanium chloride
Molecular FormulaC9H18ClNO2
InChI KeyRRHXZLALVWBDKH-UHFFFAOYSA-M
SMILES[Cl-].CC(=C)C(=O)OCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (Titration ex Chloride)≥66.5%
Assay (HPLC)≥67.0%
Refractive Index1.4660-1.4750 @ 20°C
2-(Methacryloyloxy)ethyltrimethylammonium chloride is a zirconium, aluminum and organic surface treated multipurpose rutile titanium dioxide pigment, it has narrow-size material distribution, good whiteness and high weather resistance. It is widely used in coating, plastics, paints, rubber and ink production, leather, cosmetics, soaps, plastics and in paper industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methacryloyloxy)ethyltrimethylammonium chloride is a zirconium, aluminum and organic surface treated multipurpose rutile titanium dioxide pigment, it has narrow-size material distribution, good whiteness and high weather resistance. It is widely used in coating, plastics, paints, rubber and ink production, leather, cosmetics, soaps, plastics and in paper industry.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. M Kobayashi.; M Terada.; Y Terayama.; M Kikuchi. Direct Synthesis of Well-Defined Poly[{2-(methacryloyloxy)ethyl}trimethylammonium chloride] Brush via Surface-Initiated Atom Transfer Radical Polymerization in Fluoroalcohol. Macromolecules. 2010, 43,(20), 8409-8415.
  2. D Connolly.; B Paull. High-performance separation of small inorganic anions on a methacrylate-based polymer monolith grafted with [2(methacryloyloxy)ethyl] trimethylammonium chloride. Journal of separation science. 2009, 32,(15-16), 2653-2658.