3-Phthalimidopropionic acid, 98%
3-Phthalimidopropionic acid, 98%
3-Phthalimidopropionic acid, 98%
Thermo Scientific Chemicals

3-Phthalimidopropionic acid, 98%

CAS: 3339-73-9 | C11H9NO4 | 219.196 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L13535.06
also known as L13535-06
Price (EUR)
87,30
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
87,30
Each
Chemical Identifiers
CAS3339-73-9
IUPAC Name3-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid
Molecular FormulaC11H9NO4
InChI KeyDXXHRZUOTPMGEH-UHFFFAOYSA-N
SMILESOC(=O)CCN1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)148.0-155.0?C
Appearance (Color)White
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only