Cinnamyl anthranilate, 99%
Thermo Scientific Chemicals

Cinnamyl anthranilate, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L13659.14
also known as L13659-14
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS87-29-6
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)62.5-66.5?C
FormCrystals or powder or crystalline powder or granules
Appearance (Color)Cream to pale brown
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.