3-(2-Aminoethylamino)propyltrimethoxysilane, 96%
3-(2-Aminoethylamino)propyltrimethoxysilane, 96%
3-(2-Aminoethylamino)propyltrimethoxysilane, 96%
Thermo Scientific Chemicals

3-(2-Aminoethylamino)propyltrimethoxysilane, 96%

CAS: 1760-24-3 | C8H22N2O3Si | 222.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number L14043.22
also known as L14043-22
Price (EUR)
57,80
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
57,80
Each
Chemical Identifiers
CAS1760-24-3
IUPAC Namehydrogen (2-aminoethyl)[3-(trimethoxysilyl)propyl]amine chloride
Molecular FormulaC8H23ClN2O3Si
InChI KeySZBIXNNLIKDOEJ-UHFFFAOYSA-N
SMILES[H+].[Cl-].CO[Si](CCCNCCN)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥95.0%
Identification (FTIR)Conforms
Refractive Index1.4415-1.4475 @ 20?C
3-(2-Aminoethylamino) propyltrimethoxysilane acts as a silane coupling agent. It is useful for chemical research. It is also employed as an adhesives and sealant chemical. Further, it is used as a paint additive and coating additive. In addition to this, it plays an important role in the production of petroleum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Aminoethylamino) propyltrimethoxysilane acts as a silane coupling agent. It is useful for chemical research. It is also employed as an adhesives and sealant chemical. Further, it is used as a paint additive and coating additive. In addition to this, it plays an important role in the production of petroleum.

Solubility
Miscible with toluene.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and water.
RUO – Research Use Only

General References:

  1. Peng, H.; Zhang, N.; He, M.; Chen, B.; Hu, B. Simultaneous speciation analysis of inorganic arsenic, chromium and selenium in environmental waters by 3-(2-aminoethylamino) propyltrimethoxysilane modified multi-wall carbon nanotubes packed microcolumn solid phase extraction and ICP-MS. Talanta 2015, 131, 266-272.
  2. Semiletov, A. M.; Chirkunov, A. A.; Kuznetsov, Y. I.; Andreeva, N. P. Improving steel passivation with aqueous solutions of [3-(2-Aminoethylamino)propyl]trimethoxysilane. Russ. J. Phys. Chem. A 2015, 89 (12), 2271-2277.