Triphenylphosphine hydrobromide, 97%
Triphenylphosphine hydrobromide, 97%
Triphenylphosphine hydrobromide, 97%
Triphenylphosphine hydrobromide, 97%
Thermo Scientific Chemicals

Triphenylphosphine hydrobromide, 97%

CAS: 6399-81-1 | C18H16BrP | 343.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L14290.14
also known as L14290-14
Price (EUR)
82,80
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
82,80
Each
Add to cart
Chemical Identifiers
CAS6399-81-1
IUPAC Nametriphenylphosphane hydrobromide
Molecular FormulaC18H16BrP
InChI KeyCMSYDJVRTHCWFP-UHFFFAOYSA-N
SMILESBr.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Bromide)≥96.0 to ≤104.0%
Identification (FTIR)Conforms
Triphenylphosphine hydrobromide is used as mild source of anhydrous HBr; catalyst for formation of THP ethers from tertiary alcohols; preparation of phosphonium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylphosphine hydrobromide is used as mild source of anhydrous HBr; catalyst for formation of THP ethers from tertiary alcohols; preparation of phosphonium salts.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Mani Ramanathan, Duen-Ren Hou. Cleavage of benzyl ethers by triphenylphosphine hydrobromide. Tetrahedron Letters. 2010, 51, (47), 6143-6145.
  2. C. C. Tung; A. J. Speziale. Reaction of Amides and Esters of α,β-Dibromopropionic Acids with Triphenylphosphine. J. Org. Chem. 1963, 28, (6), 1521-1523.