3,4,9,10-Perylenetetracarboxylic dianhydride, 98%
3,4,9,10-Perylenetetracarboxylic dianhydride, 98%
3,4,9,10-Perylenetetracarboxylic dianhydride, 98%
Thermo Scientific Chemicals

3,4,9,10-Perylenetetracarboxylic dianhydride, 98%

CAS: 128-69-8 | C24H8O6 | 392.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L14370.18
also known as L14370-18
Price (EUR)
76,80
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
76,80
Each
Chemical Identifiers
CAS128-69-8
IUPAC Name7,18-dioxaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Molecular FormulaC24H8O6
InChI KeyCLYVDMAATCIVBF-UHFFFAOYSA-N
SMILESO=C1OC(=O)C2=CC=C3C4=CC=C5C(=O)OC(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red
FormPowder or crystals or crystalline powder
Assay (unspecified)≥97.5%
Elemental AnalysisCarbon: 71.64 - 75.31% (Theory 73.48%) (non-U.S. specification)
Identification (FTIR)Conforms (non-U.S. specification)
3,4,9,10-Perylenetetracarboxylic dianhydride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is mainly used in air dried and baking enamels, epoxy, polyurethane and polyester based paints. It is also used in printing inks for bank and security documents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,9,10-Perylenetetracarboxylic dianhydride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is mainly used in air dried and baking enamels, epoxy, polyurethane and polyester based paints. It is also used in printing inks for bank and security documents.

Solubility
Soluble in water (0.87g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. It is hygroscopic in nature.
RUO – Research Use Only

General References:

  1. William E. Ford.; Prashant V. Kamatn. William E. Ford, Prashant V. Kamat. J. Phys. Chem. 1987, 91 (25), 6373-6380.
  2. V. Bulović.; P.E. Burrows.; S.R. Forrest.; J.A. Croninb.; M.E. Thompsonb. Study of localized and extended excitons in 3,4,9,10-perylenetetracarboxylic dianhydride (PTCDA) I. Spectroscopic properties of thin films and solutions. Chemical Physics. 1996, 210 (1-2), 1-12.